Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMP3 Antibody (32E9), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB162121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162121 Supplier Novus Biologicals Supplier No. NBP259475V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IMP3 Monoclonal antibody specifically detects IMP3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen IMP3
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 32000000000
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias BRMS2mitochondrial ribosomal protein S4, C15orf12, chromosome 15 open reading frame 12, FLJ10968, IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast), MRPS4DKFZp586L0118, U3 small nucleolar ribonucleoprotein protein IMP3, U3 snoRNP protein 3 homolog, U3 snoRNP protein IMP3
Host Species Mouse
Immunogen Recombinant human IMP3 (1-184aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 55272
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.