Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP275542 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275542 Supplier Novus Biologicals Supplier No. NBP275542

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

IMP3 Polyclonal specifically detects IMP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IMP3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:400
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias BRMS2mitochondrial ribosomal protein S4, C15orf12, chromosome 15 open reading frame 12, FLJ10968, IMP3, U3 small nucleolar ribonucleoprotein, homolog (yeast), MRPS4DKFZp586L0118, U3 small nucleolar ribonucleoprotein protein IMP3, U3 snoRNP protein 3 homolog, U3 snoRNP protein IMP3
Gene Symbols IMP3
Host Species Rabbit
Immunogen Recombinant protein within Human IMP3 aa 352-579/ 579. (SwissProt: O00425 Human; SwissProt: Q9CPN8 Mouse)
Molecular Weight of Antigen 63 kDa
Purification Method Protein A purified
Quantity 100 μL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 55272.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.