Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPA1 Antibody (2G5), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB160825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160825 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160825 Supplier Novus Biologicals Supplier No. NBP259416FR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IMPA1 Monoclonal antibody specifically detects IMPA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen IMPA1
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 2G5
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.25, IMP, IMP 1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1
Host Species Mouse
Immunogen Recombinant human IMPA1 (1-277aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 3612
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.