Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPA2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | IMPA2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
IMPA2 Polyclonal specifically detects IMPA2 in Human samples. It is validated for Western Blot.Specifications
IMPA2 | |
Polyclonal | |
Rabbit | |
Lipid and Metabolism | |
EC 3.1.3.25, IMP 2, IMP.18P, IMPase 2, inosine monophosphatase 2, inositol monophosphatase 2, inositol monophosphatase 2 variant 1, inositol monophosphatase 2 variant 2, inositol(myo)-1(or 4)-monophosphatase 2, Inositol-1(or 4)-monophosphatase 2, Myo-inositol monophosphatase A2 | |
IMPA2 | |
IgG | |
32 kDa |
Western Blot | |
Unconjugated | |
RUO | |
O14732 | |
3613 | |
Synthetic peptides corresponding to IMPA2(inositol(myo)-1(or 4)-monophosphatase 2) The peptide sequence was selected from the middle region of IMPA2. Peptide sequence RGRGAFCNGQRLRVSGETDLSKALVLTEIGPKRDPATLKLFLSNMERLLH. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title