Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPACT Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291597 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291597 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291597 Supplier Novus Biologicals Supplier No. NBP142678APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IMPACT Polyclonal antibody specifically detects IMPACT in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen IMPACT
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Impact homolog (mouse), Imprinted and ancient gene protein homolog, MGC33718, protein IMPACT
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 270 and 320 of human imprinted and ancient homolog using the numbering given in entry NP_060909.1 (GeneID 55364).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 55364
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.