Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Double Padded Anti-Impact Gloves

Catalog No. p-3668007 Shop All Impacto Protective Products Products
Encompass
Click to view available options
Size:
Large
X-Large
Hand:
Left
This item is not returnable. View return policy
This item is not returnable. View return policy

Nylon material offers breathability and comfort, while suede leather provides abrasion protection and durability. 1/4 in. VEP padding in the palm extends to the edge of the hand for protection from impact and shock.

  • VEP provides impact absorbing and shock dissipation qualities, as well as absorption of higher levels of vibration
  • Fingerless style for tactile feedback and mobility
  • Suede leather offers abrasion protection & durability
  • Side padding protects from impact during automotive applications

Specifications

Color Blue, Yellow
Hand Left
Material Nylon Lycra
Product Type Anti-impact Gloves
Size Large
Finger Style Fingerless
For Use With (Application) Automobile Construction
Glove Cuff Style Open
Series 523-24
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.