Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Anti-Impact Glove Liners with Extended VEP Padding

Catalog No. p-3142806 Shop All Impacto Protective Products Products
Encompass
Click to view available options
Size:
Small
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy

Stretch polycotton fabric ensures optimum breathability and comfort. VEP padding that extends from the palm to the wrist joint provides users with additional protection.

  • 4-way stretch polycotton fabric ensures optimum breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm extends to the wrist joint for additional protection
  • Open-finger design for tactile feedback and mobility
  • Designed to be worn under work gloves
  • Offers protection from electric, manual, and power tools
  • Use with applications such as construction, assembly, manufacturing, maintenance, and warehousing

Specifications

Color Blue
Hand Left
Material Polycotton
Product Type Glove Liners
Size Small
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Style Open
Series 510-00
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.