Learn More
Impacto™ Pearl Leather Series Half-Finger Anti-Impact Gloves
Catalog No. 19031811 Shop All Impacto Protective Products Products
Encompass
Click to view available options
Size:
Medium
Large
Hand:
Left
Right
This item is not returnable. View return policy
This item is not returnable. View return policy
Nylon back with soft pearl leather palm ensures dexterity and comfort. Impact absorbing visco-elastic polymer (VEP) padding in the palm, and adjustable hook and loop wrist closure.
  • Nylon fabric back ensures breathability and comfort
  • Palm is made of soft pearl leather for dexterity and abrasion protection
  • 1/8 in. VEP padding in the palm protects from impact and shock
  • Half-finger style leaves fingers open for tactile feedback and mobility
  • Hook and loop wrist closure
  • Protection against electric, manual, and power tools
  • Ideal for applications such as driving, sports, wheelchairs, and manufacturing
Specifications
Color Black, White
Hand Left
Material Pearl Leather, Nylon
Product Type Fingerless Gloves
Size Medium
EN388 Standard EN388:2222
Glove Cuff Style Band Top
Palm Type Padded
Series 401-30
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.