Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Leather Palm Tool Grip Gloves

Catalog No. p-3668043 Shop All Impacto Protective Products Products
Encompass
Click to view available options
Size:
Large
X-Large
Hand:
Specific

Nylon gloves are constructed with contoured VEP padding to provide protection from vibration, impact, and abrasion. The grain leather palm is smooth and provides extended wear.

  • Protection from vibration, impact, and abrasion
  • Grain leather palm is soft and durable
  • Fingerless style ensures tactile feedback
  • Use for protection from hand tools and power tools

Specifications

Back Material Nylon Lycra™
Color Blue, Yellow
Hand Specific
Material Grain Leather
Product Type Tool Gloves
Size Large
Finger Style Fingerless
For Use With (Application) Protection from Hand/Power Tools
Glove Cuff Style Open
Series 501-20
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.