Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Tennis Elbow Support

Catalog No. 19031706 Shop All Impacto Protective Products Products
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Blue
Size:
Small/Medium
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-031706 Black X-Large
19-031717 Blue Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-031706 Supplier Impacto™ Supplier No. EL5002L

Impacto™ EL5002 Tennis Elbow Support helps alleviate pain caused by lateral epicondylitis, provides support and helps prevent injuries.

Impacto™ Elbow Support

  • Reduces pain from tendonitis of the elbow, a condition caused by repetitive motion of the arm such motion is common in hammering, gardening and sports
  • Padded to compress muscle and cartilage in upper forearm, aiding in rehabilitation
  • Neoprene with adjustable hook-and-loop strap

Specifications

Color Black
Closure Type Adjustable hook and loop strap
Product Type Elbow Support
Size X-Large
For Use With (Application) Sports like golf or tennis, during gardening or while using tools
Hand Ambidextrous
Material Neoprene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.