Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Impactosol Flat GEL Insoles

Catalog No. p-7115577 Shop All Impacto Protective Products Products
Click to view available options
Size:
Men: 11 to 13, Women: 13 to 14
Men: 14 to 16
Men: 2 to 4, Women: 4 to 6
Men: 5 to 7, Women: 7 to 9
Men: 8 to 10, Women: 10 to 12

Impacto™ Impactosol Flat Gel Insoles help relieve foot fatigue and are ideal for long term wear

Impacto™ IMPACTOSOL Anti-Fatigue Insoles

  • Made with VEP gel with Poron foam top layer provides cushioning and even distribution of weight to protect pressure points
  • Anti-bacterial
  • For use with most types of footwear including low cut footwear
  • Can be trimmed for a perfect fit

Specifications

Color Blue
Length (English) 8 in.
Length (Metric) 20.32 cm
Material Gel
Size Men: 2 to 4, Women: 4 to 6
Style Open cell structure full flat insoles
Gender Unisex
Product Type Impactosol Insoles
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.