Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPDH1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152933 Supplier Novus Biologicals Supplier No. NBP152933
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IMPDH1 Polyclonal specifically detects IMPDH1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen IMPDH1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P20839
Gene Alias DKFZp781N0678, EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 1, IMP (inosine monophosphate) dehydrogenase 1, IMP dehydrogenase 1, IMPD, IMPD 1, IMPD1, IMPDH 1, IMPDH-I, inosine-5'-monophosphate dehydrogenase 1, LCA11RP10, retinitis pigmentosa 10 (autosomal dominant), sWSS2608
Gene Symbols IMPDH1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to IMPDH1(IMP (inosine monophosphate) dehydrogenase 1) The peptide sequence was selected from the middle region of IMPDH1. Peptide sequence KKGKLPIVNDCDELVAIIARTDLKKNRDYPLASKDSQKQLLCGAAVGTRE.
Molecular Weight of Antigen 62 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3614
Test Specificity Expected to cross react based on sequence identity: Yeast: 79%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.