Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166086 20 μg
NB166085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166086 Supplier Novus Biologicals Supplier No. NBP31549520UL

Rabbit Polyclonal Antibody

IMPDH1 Polyclonal antibody specifically detects IMPDH1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen IMPDH1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias DKFZp781N0678, EC 1.1.1.205, IMP (inosine 5'-monophosphate) dehydrogenase 1, IMP (inosine monophosphate) dehydrogenase 1, IMP dehydrogenase 1, IMPD, IMPD 1, IMPD1, IMPDH 1, IMPDH-I, inosine-5'-monophosphate dehydrogenase 1, LCA11RP10, retinitis pigmentosa 10 (autosomal dominant), sWSS2608
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 434-563 of human IMPDH1 (NP_899066.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3614
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.