Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IMPG2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP254954 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP254954 100 μL
NB404073 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP254954 Supplier Novus Biologicals Supplier No. NBP254954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IMPG2 Polyclonal antibody specifically detects IMPG2 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).

Specifications

Antigen IMPG2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias interphotoreceptor matrix proteoglycan 2, interphotoreceptor matrix proteoglycan 200, Interphotoreceptor matrix proteoglycan of 200 kDa, IPM 200, IPM200SPACRCAN, RP56, Sialoprotein associated with cones and rods proteoglycan, Spacrcan
Gene Symbols IMPG2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FDGGLGSGSGQKVDLITWPWSETSSEKSAEPLSKPWLEDDDSLLPAEIEDKKLVLVDKMDSTDQISKHSKYEHDDRSTHFPEEEPLSGPAVPIFADTAAESASLTLPKHISEVPGVDDYSVTKA
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 50939
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Monkey
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.