Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$336.16 - $678.13
Specifications
| Antigen | IMPG2 |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IMPG2 Polyclonal antibody specifically detects IMPG2 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).Specifications
| IMPG2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| RUO | |
| Human, Monkey | |
| interphotoreceptor matrix proteoglycan 2, interphotoreceptor matrix proteoglycan 200, Interphotoreceptor matrix proteoglycan of 200 kDa, IPM 200, IPM200SPACRCAN, RP56, Sialoprotein associated with cones and rods proteoglycan, Spacrcan | |
| IMPG2 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Rabbit | |
| Vision | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 50939 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FDGGLGSGSGQKVDLITWPWSETSSEKSAEPLSKPWLEDDDSLLPAEIEDKKLVLVDKMDSTDQISKHSKYEHDDRSTHFPEEEPLSGPAVPIFADTAAESASLTLPKHISEVPGVDDYSVTKA | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title