Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP162656
Description
IMPG2 Polyclonal specifically detects IMPG2 in Human samples. It is validated for Western Blot.Specifications
IMPG2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
interphotoreceptor matrix proteoglycan 2, interphotoreceptor matrix proteoglycan 200, Interphotoreceptor matrix proteoglycan of 200 kDa, IPM 200, IPM200SPACRCAN, RP56, Sialoprotein associated with cones and rods proteoglycan, Spacrcan | |
Rabbit | |
138 kDa | |
100 μL | |
Vision | |
50939 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9BZV3 | |
IMPG2 | |
Synthetic peptides corresponding to IMPG2(interphotoreceptor matrix proteoglycan 2) The peptide sequence was selected from the C terminal of IMPG2. Peptide sequence VVFFSLRVTNMMFSEDLFNKNSLEYKALEQRFLELLVPYLQSNLTGFQNL. | |
Affinity purified | |
RUO | |
Primary | |
Zebrafish: 86%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction