Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IMPG2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | IMPG2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
IMPG2 Polyclonal specifically detects IMPG2 in Human samples. It is validated for Western Blot.Specifications
IMPG2 | |
Polyclonal | |
Rabbit | |
Vision | |
interphotoreceptor matrix proteoglycan 2, interphotoreceptor matrix proteoglycan 200, Interphotoreceptor matrix proteoglycan of 200 kDa, IPM 200, IPM200SPACRCAN, RP56, Sialoprotein associated with cones and rods proteoglycan, Spacrcan | |
IMPG2 | |
IgG | |
138 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q9BZV3 | |
50939 | |
Synthetic peptides corresponding to IMPG2(interphotoreceptor matrix proteoglycan 2) The peptide sequence was selected from the C terminal of IMPG2. Peptide sequence VVFFSLRVTNMMFSEDLFNKNSLEYKALEQRFLELLVPYLQSNLTGFQNL. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title