Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin 11 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056484 0.1 mg
NB10056484T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056484 Supplier Novus Biologicals Supplier No. NB10056484
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Importin 11 Polyclonal specifically detects Importin 11 in Bovine, Canine, Human, Mouse, Primate, Rat samples. It is validated for Western Blot.

Specifications

Antigen Importin 11
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. Q9UI26
Gene Alias imp11, importin 11, Ran binding protein 11, Ran-binding protein 11, RANBP11, RanBP11importin-11, SLRN
Gene Symbols IPO11
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human Importin-11 was used as the immunogen (NP_001128251).
Molecular Weight of Antigen 112 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51194
Test Specificity Bovine, Chimpanzee, Dog, Human, Mouse, Rat, Rhesus Monkey. Predicted cross-reactivity based on sequence identity: Xenopus (94%).
Target Species Bovine, Mouse, Canine, Human, Mouse, Primate, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.