Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Importin 11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$181.90 - $402.00
Specifications
Antigen | Importin 11 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056484T
![]() |
Novus Biologicals
NB10056484SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056484
![]() |
Novus Biologicals
NB10056484 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
Importin 11 Polyclonal specifically detects Importin 11 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.Specifications
Importin 11 | |
Western Blot 0.5-2 ug/ml | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat, Bovine, Canine, Primate | |
Q9UI26 | |
51194 | |
A portion of amino acids 1-50 of human Importin-11 was used as the immunogen (NP_001128251). | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
112 kDa |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
imp11, importin 11, Ran binding protein 11, Ran-binding protein 11, RANBP11, RanBP11importin-11, SLRN | |
IPO11 | |
IgG | |
Affinity Purified | |
Bovine, Chimpanzee, Dog, Human, Mouse, Rat, Rhesus Monkey. Predicted cross-reactivity based on sequence identity: Xenopus (94%). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title