Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin-7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227150 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227150 0.1 mg
NBP227150SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227150 Supplier Novus Biologicals Supplier No. NBP227150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Importin-7 Polyclonal specifically detects Importin-7 in Human, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, In vitro assay.

Specifications

Antigen Importin-7
Applications Western Blot, Immunohistochemistry (Paraffin), In vitro Assay
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3ug/ml∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml, In vitro assay reported in scientific literature (PMID 24184208)
Formulation PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias FLJ14581, Imp7MGC138673, importin 7, RAN binding protein 7, Ran-binding protein 7, RanBP7, RANBP7importin-7
Gene Symbols IPO7
Host Species Rabbit
Immunogen A portion of amino acids 1-76 of human IOP7 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10527
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat, 88% homologous in xenopus and 82% homologous in pufferfish. Primate reactivity reported in scientific literature (PMID: 24100013).
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.