Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin-7 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 2 publications

$181.90 - $402.00

Specifications

Antigen Importin-7
Concentration 0.5 mg/mL
Dilution Western Blot 1-3ug/ml∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml, In vitro assay reported in scientific literature (PMID 24184208)
Applications Western Blot, Immunohistochemistry (Paraffin), In vitro Assay
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227150SS
SDP
View Documents
Novus Biologicals
NBP227150SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227150
SDP
View Documents
Novus Biologicals
NBP227150
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Importin-7 Polyclonal specifically detects Importin-7 in Human, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, In vitro assay.
Specifications

Specifications

Importin-7
Western Blot 1-3ug/ml∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml, In vitro assay reported in scientific literature (PMID 24184208)
Polyclonal
Purified
RUO
PBS and 0.05% BSA with 0.05% Sodium Azide
10527
A portion of amino acids 1-76 of human IOP7 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Western Blot, Immunohistochemistry (Paraffin), In vitro Assay
Unconjugated
Rabbit
Human, Primate
FLJ14581, Imp7MGC138673, importin 7, RAN binding protein 7, Ran-binding protein 7, RanBP7, RANBP7importin-7
IPO7
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat, 88% homologous in xenopus and 82% homologous in pufferfish. Primate reactivity reported in scientific literature (PMID: 24100013).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.