Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin-7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146168 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146168 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146168 Supplier Novus Biologicals Supplier No. NBP146168
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Importin-7 Polyclonal specifically detects Importin-7 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen Importin-7
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 5-15 ug/mg lysate, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Alias FLJ14581, Imp7MGC138673, importin 7, RAN binding protein 7, Ran-binding protein 7, RanBP7, RANBP7importin-7
Gene Symbols IPO7
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 200 and 250 of human Importin 7 using the numbering given in entry NP_006382.1 (GeneID 10527).
Molecular Weight of Antigen 120 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10527
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.