Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin-8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224751 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224751 0.1 mg
NBP224751SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224751 Supplier Novus Biologicals Supplier No. NBP224751
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Importin-8 Polyclonal specifically detects Importin-8 in Human, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen Importin-8
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:2000, Immunoprecipitation reported in scientific literature (PMID 19131589)
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O15397
Gene Alias FLJ21954, FLJ26580, Imp8, importin 8, importin-8, RAN binding protein 8, Ran-binding protein 8, RanBP8, RANBP8imp8
Gene Symbols IPO8
Host Species Rabbit
Immunogen A portion of amino acids 850-900 of human Importin-8 was used as immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10526
Test Specificity The amino acid sequence used as immunogen for the POU3F2 antibody is 100% homologous in human, horse, pig, mouse, cattle, rat and dog. Alternative initiation produces three different forms of this protein (approximately 47, 29 and 27 kD).
Target Species Human, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.