Learn More
Importin-9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Importin-9 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | DKFZp761M1547, FLJ10402, Imp9IMP9, importin 9, importin-9, KIAA1192, Ran-binding protein 9, RANBP9 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 827-1041 of human Importin-9 (NP_060555.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.