Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin alpha 2/KPNA2 Rabbit anti-Human, Mouse, Rat, Clone: 9T3V8, Novus Biologicals™
SDP

Catalog No. NB168151 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168151 100 μg
NB168152 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB168151 Supplier Novus Biologicals Supplier No. NBP316530100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Importin alpha 2/KPNA2 Monoclonal antibody specifically detects Importin alpha 2/KPNA2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Importin alpha 2/KPNA2
Applications Western Blot
Classification Monoclonal
Clone 9T3V8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias importin alpha 1, importin-alpha-P1, IPOA1, karyopherin alpha 2 (RAG cohort 1, importin alpha 1), Karyopherin subunit alpha-2, pendulin, QIP2importin alpha 2, RAG cohort 1, RAG cohort protein 1, RCH1importin subunit alpha-2, SRP1, SRP1alpha, SRP1-alpha
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Importin alpha 2/KPNA2 (P52292).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3838
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.