Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Importin alpha 3/KPNA4 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB358498 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358498 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358498 Supplier Novus Biologicals Supplier No. NBP176300PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Importin alpha 3/KPNA4 Polyclonal antibody specifically detects Importin alpha 3/KPNA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Importin alpha 3/KPNA4
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias importin alpha 3, Importin alpha Q1, importin subunit alpha-4, importin-alpha-Q1, IPOA3, karyopherin alpha 4 (importin alpha 3), MGC12217, MGC26703, Qip1, QIP1Karyopherin subunit alpha-4, SRP3
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human KPNA4. The immunogen is located within amino acids 30 - 80 of KPNA4. Amino Acid Squence: RNVPQEDICEDS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3840
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.