Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Chemicon™ In Vitro Vascular Permeability Assay

Catalog No. ecm642 Shop All MilliporeSigma Products
Click to view available options
Quantity:
24 Assays
96 Assays

Catalog No. ECM642

Supplier: MilliporeSigma™ ECM642

Only null left
Add to Cart
Add to Cart

For evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store kit materials at 2-8°C; use within 4 months from date of receipt
Description In Vitro Vascular Permeability Assay
Detection Method Fluorescence
For Use With (Application) Evaluating effects of chemicals and drug compounds on endothelial cell adsorption, transport and permeability
Includes Black plate, 96-well opaque/permeability insert assembly collagen-coated, receiver tray, FITC-dextran solution, cell stain
Quantity 96 Assays
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.