Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INAVA Antibody (OTI6A6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB288436 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288436 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288436 Supplier Novus Biologicals Supplier No. NBP272021APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

INAVA Monoclonal antibody specifically detects INAVA in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen INAVA
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6A6
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias C1orf106, chromosome 1 open reading frame 106, hypothetical protein LOC55765
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-270 of human C1ORF106(NP_060735) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55765
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.