Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ INCA-6
GREENER_CHOICE

Catalog No. p-5545830
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of calcineurin-substrate association

INCA-6 is an inhibitor of interaction between calcineurin and its substrate nuclear factor of activated T cells (NFAT); blocks at the substrate recognition site but not at the catalytic site (Kd = 0.8 mM). Inhibits NFAT dephosphorylation and nuclear import. Also prevents induction of cytokine mRNAs that are downstream targets of NFAT.

Specifications

Quantity 10 mg
Product Type INCA-6
Purity >98%
Molecular Weight (g/mol) 284.31
Inhibitors INCA-6

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.