Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

INCENP Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095258 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095258 100 μL
NB095259 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095258 Supplier Novus Biologicals Supplier No. NBP305157100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

INCENP Polyclonal specifically detects INCENP in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen INCENP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias binds and activates aurora-B and -C in vivo and in vitro, chromosomal passenger protein, FLJ31633, inner centromere protein, inner centromere protein antigens (135kD, 155kD), inner centromere protein antigens 135/155kDa, inner centromere protein INCENP, MGC111393
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 785-914 of human INCENP (NP_064623.2).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3619
Target Species Human, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.