Learn More
Azure Biosystems Incubation Tray
SDP
Catalog No. p-200054227
Click to view available options
Dimensions (L x W):
11 x 9 cm
15.5 x 10.5 cm
7 x 5 cm
9 x 6 cm
Tray with clear lid and bottom for staining and washing gels and membranes.
  • Four sizes available
  • These Western blot incubation boxes are suitable for staining and washing gels and membranes
  • Western blot boxes have smooth interiors to avoid damage to gels and membranes and lids to protect them from dust and debris
Specifications
Color Clear
Type Blot Processing Tray
Quantity 2 Trays
Dimensions (L x W) 15.5 x 10.5 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.