Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indian Hedgehog/Ihh Antibody (9L16), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP222020IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222020IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222020IR Supplier Novus Biologicals Supplier No. NBP222020IR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Indian Hedgehog/Ihh
Applications Western Blot, ELISA
Classification Monoclonal
Clone 9L16
Conjugate DyLight 755
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein
Gene Symbols IHH
Host Species Rat
Immunogen Human recombinant Indian Hedgehog/Ihh (N-terminus)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 3549
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.