Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Indisulam
GREENER_CHOICE

Catalog No. p-7166798
Click to view available options
Quantity:
5 mg
25 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Molecular glue; pre-mRNA splicing modulator

Indisulam is an acts as a molecular glue to induce proteosomal degradation of mRNA splicing factor RBM39 (also designated CAPERα, HCC1, FSAP59, and RNPC2), via binding to DCAF15. Acts as a pre-mRNA splicing modulator (SPLAMs; splicing inhibitor sulfonamides), causes aberrant pre-mRNA splicing. Suppresses proliferation of cancer cell lines. Reduces viability of HCT-116 cells (IC50 = 0.56 μM). Induces cell cycle arrest in the G1 phase in cancer cell lines. Also a high affinity carbonic anhydrase isozyme XII (hCA XII) inhibitor (Ki = 3.0-5.7 nM).

Specifications

Chemical Name or Material N1-(3-Chloro-1 H-indol-7-yl)-1,4-benzenedisulfonamide
CAS 165668-41-7
Quantity 5 mg
Target Ubiquitin Ligase (E3) Inhibitors
Molecular Formula C14H12ClN3O4S2
Formula Weight 385.84
Product Type pre-mRNA Splicing Modulator
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.