Learn More
Indium(III) chloride hydrate, 99.99% (metals basis)
Description
Indium(III) chloride hydrate is used for mild and selective cleavage of tert-butyldimethylsilyl ethers. It is a Lewis acid, and serves as a catalyst for many organic transformations. It also catalyzes the conversion of aldehydes to gem diacetates by chemoselective process. Lithium indium hydride, an in situ reducing agent, can be prepared from indium trichloride. It catalyzes reactions such as Friedel-Crafts acylations and Diels-Alder reactions. It also catalyzes Knoevenagel condensation between the barbituric acid and the aldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline |
| Quantity | 10 g |
| UN Number | UN3260 |
| Merck Index | 14,4958 |
| Solubility Information | Soluble in water,diethyl ether,and THF. |
| Formula Weight | 221.18 (Anhydrous) |
| Percent Purity | 99.99% |
| Sensitivity | Hygroscopic |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 233-043-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.