Learn More
Indium(III) hydroxide, 99.8% (metals basis)
Description
Indium(III) hydroxide is used as a precursor to indium(III)oxide. It is also applied in batteries, glasses, ceramics, chemical reagents and semiconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 150°C (in -H2O) |
| Physical Form | Powder |
| Quantity | 50 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 165.84 |
| Percent Purity | 99.8% |
| Assay | (metals basis) |
| Chemical Name or Material | Indium(III) hydroxide |
Safety and Handling
| Hazard Category | H315-H319 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 243-947-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.