Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoleamine 2,3-dioxygenase/IDO Antibody (700838), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB303104 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303104 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303104 Supplier Novus Biologicals Supplier No. FAB6030A

Mouse Monoclonal Antibody

Indoleamine 2,3-dioxygenase/IDO Monoclonal antibody specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Indoleamine 2,3-dioxygenase/IDO
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 700838
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
Host Species Mouse
Immunogen E. coli-derived recombinant human Indoleamine 2,3-dioxygenase/IDO, Ala2-Gly403, Accession # P14902
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 3620
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.