Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoleamine 2,3-dioxygenase/IDO Antibody (998736), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB295976 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295976 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295976 Supplier Novus Biologicals Supplier No. FAB60302E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Indoleamine 2,3-dioxygenase/IDO Monoclonal specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot.

Specifications

Antigen Indoleamine 2,3-dioxygenase/IDO
Applications CyTOF, Flow Cytometry (Intracellular Staining), Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 998736
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
Host Species Mouse
Immunogen E. coli-derived human Indoleamine 2,3-dioxygenase/IDO, Ala2-Gly403, Accession # P14902
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 3620
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.