Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
INF 39
Click to view available options
Quantity:
10 mg
Description
INF 39 is an irreversible NLRP3 inhibitor (exhibits 52% inhibition of NLRP3 ATPase at 100 μM). Inhibits IL-1β release, caspase activation and pyroptosis in THP-1 cells. Reduces inflammation in a rat colitis model. Exhibits low cytotoxicity.
Specifications
Specifications
Chemical Name or Material | Ethyl 2-(2-chlorobenzyl)acrylate |
CAS | 866028-26-4 |
Assay Percent Range | >98% |
Quantity | 10 mg |
Molecular Formula | C12H13ClO2 |
Formula Weight | Observed MW: 224.68 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction