Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Infectious Bronchitis Virus Mouse anti-Virus, Clone: IB95, Novus Biologicals™
SDP

Catalog No. NB093865 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093865 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB093865 Supplier Novus Biologicals Supplier No. NBP2311020.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Infectious Bronchitis Virus Monoclonal specifically detects Infectious Bronchitis Virus in Virus samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Sandwich ELISA.

Specifications

Antigen Infectious Bronchitis Virus
Applications Western Blot, ELISA
Classification Monoclonal
Clone IB95
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with semi-purified inactivated IB virus of Massachusetts type from infected chicken embryos.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.