Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H10N8 Hemagglutinin Antibody (137) - (A/Jiangxi-Donghu/346/2013)
SDP

Catalog No. NB022139 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB022139 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB022139 Supplier Novus Biologicals Supplier No. NBP290341

Rabbit Monoclonal Antibody

Influenza A H10N8 Hemagglutinin Monoclonal specifically detects Influenza A H10N8 Hemagglutinin in Virus samples. It is validated for ELISA.

Specifications

Antigen Influenza A H10N8 Hemagglutinin
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 137
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant H10N8 Influenza A H10N8 Hemagglutinin (A/Jiangxi-Donghu/346/2013) (EPI497477; Met1-Asp524).
Purification Method Protein A purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Influenza A H10N8 (A/Jiangxi-Donghu/346/2013) Hemagglutinin/HA
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.