Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Antibody - (A/California/14/2009), Novus Biologicals™
SDP

Catalog No. NB018479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018479 0.025 mg
NBP130320 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018479 Supplier Novus Biologicals Supplier No. NBP130320SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Influenza A H1N1 Polyclonal antibody specifically detects Influenza A H1N1 in Virus samples. It is validated for Western Blot.

Specifications

Antigen Influenza A H1N1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Alias Flu
Gene Symbols HA
Host Species Rabbit
Immunogen A/California/14/2009 (H1N1) virions were used as an immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 956529
Test Specificity This antibody detects the following H1N1 proteins: Hemagglutinin (HA) ∼75 kDa, Neuraminidase monomer (NA) ∼55 kDa, Matrix (M) ∼26 kDa and Non-structural Protein monomer (NS) ∼26-27 kDa. Reacts with A/New Caledonia/20/99, A/Brisbane/10/2007, A/California/14/2009 and likely reacts with most other isolates.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.