Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Hemagglutinin Antibody (10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB351248 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351248 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351248 Supplier Novus Biologicals Supplier No. NBP306578MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza virusN1 samples. It is validated for Western Blot

Specifications

Antigen Influenza A H1N1 Hemagglutinin
Applications Western Blot
Classification Monoclonal
Clone 10
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H1N1 Hemagglutinin (A/California/04/2009)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus N1
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.