Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Hemagglutinin Antibody - (A/USSR/90/1977) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017433 0.1 mL
NB10062125 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017433 Supplier Novus Biologicals Supplier No. NB100621250.1ML
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Influenza A H1N1 Hemagglutinin Polyclonal specifically detects Influenza A H1N1 Hemagglutinin in Influenza A virus H1N1 samples. It is validated for Haemagglutination, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Influenza A H1N1
Applications Hemagglutination Assay, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Haemagglutination
Formulation PBS with 0.09% Sodium Azide
Gene Alias HA, Hemagglutinin
Host Species Goat
Immunogen Influenza A, strain USSR (H1N1)
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 956529
Test Specificity Specific for Influenza A virus H1N1 glycoprotein and does not react with Influenza B, RSV, Para 1-3 or Adenovirus. It does not react with HEp-2 cells but may react with some chicken cellular proteins.
Target Species Influenza A virus H1N1, Influenza virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.