Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Hemagglutinin Antibody (IV.C102), DyLight 405 - (A/USSR/90/77), Novus Biologicals™
SDP

Catalog No. NB10065047V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065047V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065047V Supplier Novus Biologicals Supplier No. NB10065047V

Mouse Monoclonal Antibody

Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza A Virus H1N1 samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).

Specifications

Antigen Influenza A H1N1 Hemagglutinin
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone IV.C102
Conjugate DyLight 405
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Sandwich ELISA
Gene Alias HA, HA1, HA2, Hemagglutinin, Influenza A Virus Hemagglutinin
Gene Symbols HA
Host Species Mouse
Immunogen Influenza A/USSR/90/77
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-65047 is specific for the haemagglutinin of the Influenza A H1N1 serotype and does not react with H2N2, H3N2, Influenza B, RSV, Parainfluenza I, II or III, Adenovirus, mumps or measle antigens.
Target Species Influenza A virus H1N1, Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.