Learn More
Description
Specifications
Specifications
| Antigen | Influenza A H1N1 Hemagglutinin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1-1 ug/ml |
| Formulation | 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide |
| Gene Alias | HA, HA1, HA2, Hemagglutinin, Influenza A Virus Hemagglutinin |
| Host Species | Rabbit |
| Immunogen | Within the range of amino acids 320-362 of human Influenza A H1N1 NA was used as the immunogen. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
