Learn More
Description
Specifications
Specifications
| Antigen | Influenza A H1N1 Hemagglutinin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | HA, HA1, HA2, Hemagglutinin, Influenza A Virus Hemagglutinin |
| Host Species | Rabbit |
| Immunogen | Within the range of amino acids 35-58 of Influenza A H1N1 NA (swine flu) were used as the immunogen. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
