Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Hemagglutinin Antibody, Novus Biologicals™
SDP

Catalog No. NBP224955SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224955SS 0.025 mg
NBP224955 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224955SS Supplier Novus Biologicals Supplier No. NBP224955SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Influenza A H1N1 Hemagglutinin Polyclonal specifically detects Influenza A H1N1 Hemagglutinin in Influenza A Virus H1N1 samples. It is validated for Western Blot.

Specifications

Antigen Influenza A H1N1 Hemagglutinin
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Alias HA, HA1, HA2, Hemagglutinin, Influenza A Virus Hemagglutinin
Host Species Rabbit
Immunogen Within the range of amino acids 35-58 of Influenza A H1N1 NA (swine flu) were used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza A virus H1N1, Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.