Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 M1 Antibody (02), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB350959 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350959 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350959 Supplier Novus Biologicals Supplier No. NBP306633MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Influenza A H1N1 M1 Monoclonal antibody specifically detects Influenza A H1N1 M1 in Influenza virusN1 samples. It is validated for ELISA

Specifications

Antigen Influenza A H1N1 M1
Applications ELISA
Classification Monoclonal
Clone 02
Conjugate mFluor Violet 450 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H1N1 (Accession#: NP_040978.1; Ser 2-Lys 252).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus N1
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.