Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Neuraminidase Antibody (GT172) - (A/WSN/1933), Novus Biologicals™
SDP

Catalog No. NB177181 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177181 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177181 Supplier Novus Biologicals Supplier No. NBP313494
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Influenza A H1N1 Neuraminidase Monoclonal antibody specifically detects Influenza A H1N1 Neuraminidase in Influenza Virus N1 samples. It is validated for Western Blot

Specifications

Antigen Influenza A H1N1 Neuraminidase
Applications Western Blot
Classification Monoclonal
Clone GT172
Conjugate Unconjugated
Formulation PBS
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the Internal region of Influenza A Virus H1N1 Neuraminidase (Neuraminidase) (A/WSN/1933(H1N1). The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Influenza virus N1
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.