Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 NS2 Antibody - (A/Puerto Rico/8/1934), Novus Biologicals™
SDP

Catalog No. NBP242873 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242873 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242873 Supplier Novus Biologicals Supplier No. NBP242873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Influenza A Virus NS2 in Virus samples, and it is validated for Western Blotting

Influenza A H1N1 NS2 Polyclonal antibody specifically detects Influenza A H1N1 NS2 in Virus samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen Influenza A H1N1 NS2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence
Formulation PBS (pH 7.0) and 20% Glycerol with 0.01% Thimerosal
Gene Alias Influenza NS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of Influenza A virus NS2 (nonstructural protein) (A/Puerto Rico/8/1934(H1N1)). The exact sequence is proprietary.
Molecular Weight of Antigen 14 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.